missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoproterenol Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Isoprenaline Hydrochloride |
| CAS | 51-30-9 |
| Molecular Formula | C11 H17 N O3 . Cl H |
| Quantity | 1 g |
| Synonym | Isoprenaline hydrochloride, (1RS)-1-(3,4-Dihydroxyphenyl)-2-[(1-methylethyl)amino]ethanol hydrochloride, 1,2-Benzenediol, 4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]-, hydrochloride (1:1), 1,2-Benzenediol, 4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]-, hydrochloride (9CI), Benzyl alcohol, 3,4-dihydroxy-α-[(isopropylamino)methyl]-, hydrochloride (8CI), Protocatechuyl alcohol, α-(isopropylaminomethyl)-, hydrochloride (5CI), (±)-Isoprenaline hydrochloride, (±)-Isopropylnoradrenaline hydrochloride, (±)-Isoproterenol hydrochloride, 1-(3,4-Dihydroxyphenyl)-2-(isopropylamino)ethanol hydrochloride, 3,4-Dihydroxy-α-(isopropylaminomethyl)benzyl alcohol hydrochloride, Aerolone, Aerotrol, Asthpul, DL-Isoprenaline hydrochloride, DL-Isoproterenol hydrochloride, Euspiran, Isadrine, Isomenyl, Isoprel, Isoprenaline chloride, Isopropylarterenol hydrochloride, Isoproterenol hydrochloride, Isovon, Isuprel, Mistarel, NSC 37745, NSC 89747, Proternol, Saventrine, Suscardia, Vapo-Iso, dl-Isadrine hydrochloride, dl-Isoprenaline hydrochloride, dl-Isopropylnoradrenaline hydrochloride, dl-Isopropylnorepinephrine hydrochloride, dl-Isoproterenol hydrochloride, α-(Isopropylaminomethyl)-3,4-dihydroxybenzyl alcohol hydrochloride |
| InChI Formula | InChI=1S/C11H17NO3.ClH/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;/h3-5,7,11-15H,6H2,1-2H3;1H |
| SMILES | Cl.CC(C)NCC(O)c1ccc(O)c(O)c1 |
| IUPAC Name | 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride |
| Molecular Weight (g/mol) | 247.72 |
| Formula Weight | 247.0975 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?