missing translation for 'onlineSavingsMsg'
Learn More

Isopropyltriphenylphosphonium iodide, 98%

Product Code. 10306880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10306880 25 g Glass bottle 25g
10215380 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10306880 Supplier Thermo Scientific Acros Supplier No. 137080250
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24470-78-8 | C21H22IP | 432.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24470-78-8
Molecular Formula C21H22IP
Molecular Weight (g/mol) 432.29
MDL Number MFCD00031595
InChI Key HHBXWXJLQYJJBW-UHFFFAOYSA-M
Synonym isopropyltriphenylphosphonium iodide, phosphonium, 1-methylethyl triphenyl-, iodide, isopropyltriphenylphosphanium iodide, isopropyl triphenyl phosphonium iodide, triphenyl propan-2-yl phosphanium iodide, i-propyl triphenylphosphonium iodide, isopropyl triphenylphosphonium iodide, acmc-209gck, ksc493k7j, isopropyltriphenylphosphine iodide
PubChem CID 2723783
IUPAC Name triphenyl(propan-2-yl)phosphanium;iodide
SMILES [I-].CC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 192.0°C to 197.0°C
Color Beige-Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2CHPPh3+I-
Quantity 25 g
Formula Weight 432.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Isopropyltriphenylphosphonium iodide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Isopropyltriphenylphosphonium iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.