missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmethylphenol, TRC

Product Code. 30396749
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396749 5 g 5g
30408635 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396749 Supplier Toronto Research Chemicals Supplier No. TRCI2222805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3228-02-2
Molecular Formula C10 H14 O
Molecular Weight (g/mol) 150.22
Synonym 3-Methyl-4-(1-methylethyl)phenol, m-Cresol, 4-isopropyl- (4CI), o-Cymen-5-ol (7CI,8CI), 3-Methyl-4-(1-methylethyl)phenol, 1-Hydroxy-3-methyl-4-isopropylbenzene, 3-Methyl-4-(propan-2-yl)phenol, 3-Methyl-4-isopropylphenol, 4-Isopropyl-3-methylphenol, 4-Isopropyl-5-methylphenol, 4-Isopropyl-m-cresol, 5-Methyl-4-isopropylphenol, Biosol, Biosol (antibacterial), Frecide, NSC 62111, p-Thymol
IUPAC Name 3-methyl-4-propan-2-ylphenol
SMILES CC(C)c1ccc(O)cc1C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H14O/c1-7(2)10-5-4-9(11)6-8(10)3/h4-7,11H,1-3H3
Formula Weight 150.1045
Percent Purity >95%
Chemical Name or Material 4-Isopropyl-3-methylphenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.