missing translation for 'onlineSavingsMsg'
Learn More

Isopropylboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11384336
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11384336

Brand: Thermo Scientific Alfa Aesar L19962.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Isopropylboronic acid as a reagent is involved in copper-promoted cross-coupling, Domino Heck-Suzuki reactions, Suzuki-Miyaura type couple reactions and alkylation-hydride reduction sequence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylboronic acid as a reagent is involved in copper-promoted cross-coupling, Domino Heck-Suzuki reactions, Suzuki-Miyaura type couple reactions and alkylation-hydride reduction sequence.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80041-89-0
Molecular Formula C3H9BO2
Molecular Weight (g/mol) 87.913
MDL Number MFCD01319021
InChI Key QIPHSSYCQCBJAX-UHFFFAOYSA-N
Synonym isopropylboronic acid, propan-2-yl boronic acid, 2-propaneboronic acid, prop-2-yl boronic acid, 1-methylethyl boronic acid, boronic acid, 1-methylethyl, isopropylboric acid, pubchem7987, isopropyl boronic acid, acmc-209piu
PubChem CID 2734750
IUPAC Name propan-2-ylboronic acid
SMILES B(C(C)C)(O)O

Specifications

Quantity 1 g
Beilstein 1731883
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 87.91
Percent Purity 98%
Chemical Name or Material Isopropylboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.