missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine, TRC

Product Code. 30407646
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407646 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407646 Supplier Toronto Research Chemicals Supplier No. TRCI82397550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropylamine
CAS 75-31-0
Purity Grade Notes HPLC
Molecular Formula C3H9N
Quantity 50 g
Synonym 2-Propanamine, Isopropylamine (8CI), 1-Methylethylamine, 2-Aminopropane, 2-Isopropylamine, 2-Propylamine, Monoisopropylamine, NSC 62775, Propan-2-ylamine, sec-Propylamine
InChI Formula InChI=1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3
SMILES CC(C)N
IUPAC Name propan-2-amine
Molecular Weight (g/mol) 59.11
Formula Weight 59.0735
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.