Product Code. 10724353
Shop All Thermo Scientific Chemicals Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10724353
5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
10724353
Supplier
Thermo Scientific Acros
Supplier No.
309090050
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Specifications
CAS
400-38-4
Color
Colorless
Density
1.1000g/mL
Boiling Point
73.0°C (749.0 mmHg)
Flash Point
−8°C
Infrared Spectrum
Authentic
Assay Percent Range
98%
Packaging
Glass bottle
Molecular Formula
C5H7F3O2
Linear Formula
CF3 CO2 CH(CH3 )2
Refractive Index
1.3170 to 1.3190
MDL Number
MFCD00013566
Quantity
5 g
Specific Gravity
1.1
Synonym
isopropyl trifluoroacetate, isopropyltrifluoroacetate, isopropyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, 1-methylethyl ester, iso-propyl trifluoroacetate, trifluoroacetic acid isopropyl ester, pubchem12590, acmc-1ctsh, methylethyl 2,2,2-trifluoroacetate
InChI Key
ASAXRKSDVDALDT-UHFFFAOYSA-N
SMILES
CC(C)OC(=O)C(F)(F)F
IUPAC Name
propan-2-yl 2,2,2-trifluoroacetate
Molecular Weight (g/mol)
156.10
PubChem CID
78998
Formula Weight
156.11
Percent Purity
98%
Physical Form
Liquid
Chemical Name or Material
Isopropyl trifluoroacetate, 98%
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?