missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Tenofovir, TRC

Product Code. 30402222
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402222 2.5 mg 2.5mg
30391569 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402222 Supplier Toronto Research Chemicals Supplier No. TRCI8729302.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346597-36-1
Molecular Formula C12H20N5O4P
Molecular Weight (g/mol) 329.29
Synonym [[(1R)-2(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl] Isopropyl Phosphonic Acid
IUPAC Name [(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-propan-2-yloxyphosphinic acid
SMILES CC(C)OP(=O)(O)CO[C@H](C)Cn1cnc2c(N)ncnc12

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C12H20N5O4P/c1-8(2)21-22(18,19)7-20-9(3)4-17-6-16-10-11(13)14-5-15-12(10)17/h5-6,8-9H,4,7H2,1-3H3,(H,18,19)(H2,13,14,15)/t9-/m1/s1
Formula Weight 329.1253
Chemical Name or Material Isopropyl Tenofovir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.