missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-tartronic Acid Diethyl Ester, TRC

Product Code. 30402298
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402298 500 mg 500mg
30400352 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402298 Supplier Toronto Research Chemicals Supplier No. TRCI873570500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl-tartronic Acid Diethyl Ester
CAS 24124-04-7
Molecular Formula C10H18O5
Quantity 500 mg
Synonym Hydroxy(1-methylethyl)-propanedioic Acid Diethyl Ester, Diethyl Isopropyltartronate, Diethyl Hydroxyisopropylmalonate, NSC 157965
InChI Formula InChI=1S/C10H18O5/c1-5-14-8(11)10(13,7(3)4)9(12)15-6-2/h7,13H,5-6H2,1-4H3
SMILES CCOC(=O)C(O)(C(C)C)C(=O)OCC
IUPAC Name diethyl 2-hydroxy-2-propan-2-ylpropanedioate
Molecular Weight (g/mol) 218.25
Formula Weight 218.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.