missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl-tartronic Acid Diethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Isopropyl-tartronic Acid Diethyl Ester |
| CAS | 24124-04-7 |
| Molecular Formula | C10H18O5 |
| Quantity | 100 mg |
| Synonym | Hydroxy(1-methylethyl)-propanedioic Acid Diethyl Ester, Diethyl Isopropyltartronate, Diethyl Hydroxyisopropylmalonate, NSC 157965 |
| InChI Formula | InChI=1S/C10H18O5/c1-5-14-8(11)10(13,7(3)4)9(12)15-6-2/h7,13H,5-6H2,1-4H3 |
| SMILES | CCOC(=O)C(O)(C(C)C)C(=O)OCC |
| IUPAC Name | diethyl 2-hydroxy-2-propan-2-ylpropanedioate |
| Molecular Weight (g/mol) | 218.25 |
| Formula Weight | 218.1154 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?