missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Palmitate, Mikromol™

Product Code. 17980816
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980816 Supplier Mikromol Supplier No. MM1479.00
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Isopropyl Palmitate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl hexadecanoate
CAS 142-91-6
Analyte or Component Names Isopropyl hexadecanoate
Shipping Condition Room Temperature
Molecular Formula C19 H38 O2
Certifications/Compliance ISO 17034
Synonym Isopropyl palmitate, Hexadecanoic acid, 1-methylethyl ester, 1-Methylethyl hexadecanoate, Palmitic acid, isopropyl ester (6CI,7CI,8CI), 1-Methylethyl hexadecanoate, Crodamol IPP, Deltyl, Deltyl Prime, Emcol IP, Emerest 2316, Estol 1517, Exceparl IPP, IPP, IPP-EX, Isopal, Isopalm, Isopropyl hexadecanoate, Isopropyl palmitate, Kessco IPP, Lexol IPP, NSC 69169, Neoderm IPP, Nikkol IPP, Nikkol IPP-EX, Palmsurf IPP 98, Propal, Sinnoester PIT, Stepan D 70, Stepan IPP, Versagel MP 1600, Wickenol 111
InChI Formula InChI=1S/C19H38O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19(20)21-18(2)3/h18H,4-17H2,1-3H3
SMILES CCCCCCCCCCCCCCCC(=O)OC(C)C
IUPAC Name propan-2-yl hexadecanoate
Molecular Weight (g/mol) 298.5
Quantity 100 mg
Formula Weight 298.2872 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.