missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl myristate, 96%

Product Code. 10676504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10676504 1 kg Glass bottle 1kg
10710682 25 g Glass bottle 25g
10687191 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10676504 Supplier Thermo Scientific Acros Supplier No. 412850010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-27-0 | C17H34O2 | 270.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-27-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.45
MDL Number MFCD00008982
InChI Key AXISYYRBXTVTFY-UHFFFAOYSA-N
Synonym isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester
PubChem CID 8042
IUPAC Name propan-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Acidity 0.5mg KOH/g max.
Density 0.8500g/mL
Boiling Point 167.0°C (9.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)12COOCH(CH3)2
Refractive Index 1.4340 to 1.4360
Quantity 1 kg
Merck Index 15, 5261
Specific Gravity 0.85
Formula Weight 270.45
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Isopropyl myristate, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.