missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Hydrogen Methylphosphonate, TRC

Product Code. 30392497
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392497 Supplier Toronto Research Chemicals Supplier No. TRCI8246355G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl methylphosphonic acid
CAS 1832-54-8
Molecular Formula C4 H11 O3 P
Quantity 5 g
Synonym IMPA, Isopropyl Methylphosphonate, Isopropyl Methylphosphonic Acid, Monoisopropyl Methylphosphonate, O-Isopropyl Hydrogen Methylphosphonate, O-Isopropyl Methylphosphonic Acid, Mono(1-methylethyl) P-methyl-phosphonic Acid Ester
InChI Formula InChI=1S/C4H11O3P/c1-4(2)7-8(3,5)6/h4H,1-3H3,(H,5,6)
SMILES CC(C)OP(=O)(C)O
IUPAC Name methyl(propan-2-yloxy)phosphinic acid
Molecular Weight (g/mol) 138.1
Formula Weight 138.0446
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.