missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl glycolate, 96%

Product Code. 11429510
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429510 5 g 5g
11419510 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429510 Supplier Thermo Scientific Alfa Aesar Supplier No. H50334.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 623-61-0 | C5H10O3 | 118.132 g/mol

Isopropyl glycolate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl glycolate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-61-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00046747
InChI Key AZKIQQBSVTWCGY-UHFFFAOYSA-N
Synonym isopropyl glycolate, isopropyl 2-hydroxyacetate, isopropyl hydroxyacetate, 2-hydroxyacetic acid isopropyl ester, acetic acid, hydroxy-, 1-methylethyl ester, isopropylglycolate, acmc-209x9i, glycolic acid isopropyl ester, ksc614s5p, hydroxy-acetic acid isopropyl ester
PubChem CID 69338
IUPAC Name propan-2-yl 2-hydroxyacetate
SMILES CC(C)OC(=O)CO

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 118.13
Percent Purity 96%
Chemical Name or Material Isopropyl glycolate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.