missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl glycolate, 96%

Product Code. 11419510
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11419510

Brand: Thermo Scientific Alfa Aesar H50334.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-61-0 | C5H10O3 | 118.132 g/mol

Isopropyl glycolate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl glycolate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-61-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00046747
InChI Key AZKIQQBSVTWCGY-UHFFFAOYSA-N
Synonym isopropyl glycolate, isopropyl 2-hydroxyacetate, isopropyl hydroxyacetate, 2-hydroxyacetic acid isopropyl ester, acetic acid, hydroxy-, 1-methylethyl ester, isopropylglycolate, acmc-209x9i, glycolic acid isopropyl ester, ksc614s5p, hydroxy-acetic acid isopropyl ester
PubChem CID 69338
IUPAC Name propan-2-yl 2-hydroxyacetate
SMILES CC(C)OC(=O)CO

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 118.13
Percent Purity 96%
Chemical Name or Material Isopropyl glycolate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.