missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Diphenyl Phosphate-d7, TRC

Product Code. 30396027
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396027 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30396027 Supplier Toronto Research Chemicals Supplier No. TRCI82447210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl Diphenyl Phosphate-d7
CAS 2469626-65-9
Molecular Formula C15H10D7O4P
Quantity 10 mg
Synonym Diphenyl Isopropyl Phosphate-d7, 1-Methylethyl Diphenyl Ester Phosphoric Acid-d7
InChI Formula InChI=1S/C15H17O4P/c1-13(2)17-20(16,18-14-9-5-3-6-10-14)19-15-11-7-4-8-12-15/h3-13H,1-2H3/i1D3,2D3,13D
SMILES O=P(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC(C([2H])([2H])[2H])([2H])C([2H])([2H])[2H]
IUPAC Name diphenyl (propan-2-yl-d7) phosphate
Molecular Weight (g/mol) 299.31
Formula Weight 299.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.