missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-d7 Paraben, TRC

Product Code. 30389052
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389052 50 mg
30396188 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30389052 Supplier Toronto Research Chemicals Supplier No. TRCI87286250MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1432063-95-0
Molecular Formula C10H5D7O3
Molecular Weight (g/mol) 187.24
Synonym 4-Hydroxybenzoic Acid 1-Methylethyl-d7 Ester, 4-Hydroxybenzoic Acid Isopropyl-d7 Ester, Isopropyl-d7 4-Hydroxybenzoate, Isopropyl-d7 p-Hydroxybenzoate, p-Hydroxybenzoic Acid Isopropyl-d7 Ester
SMILES OC1=CC=C(C(OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)C=C1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H12O3/c1-7(2)13-10(12)8-3-5-9(11)6-4-8/h3-7,11H,1-2H3/i1D3,2D3,7D
Chemical Name or Material Isopropyl-d7 Paraben
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.