missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-d7-amine Hydrochloride, TRC

Product Code. 30404996
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404996 25 mg 25mg
30400882 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30404996 Supplier Toronto Research Chemicals Supplier No. TRCI82397625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl-d7-amine Hydrochloride
CAS 106658-10-0
Molecular Formula C3 2H7 H2 N . Cl H
Quantity 25 mg
Synonym 2-Propan-1,1,1,2,3,3,3-d7-amine, hydrochloride (9CI), Isopropyl-d7-amine Hydrochloride
InChI Formula InChI=1S/C3H9N.ClH/c1-3(2)4;/h3H,4H2,1-2H3;1H/i1D3,2D3,3D;
SMILES Cl.[2H]C([2H])([2H])C([2H])(N)C([2H])([2H])[2H]
IUPAC Name 1,1,1,2,3,3,3-heptadeuteriopropan-2-amine;hydrochloride
Molecular Weight (g/mol) 102.61
Formula Weight 102.0941
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.