missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl ß-D-Thiogalactopyranoside, Dioxane Free, TRC

Product Code. 30399083
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30399083 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30399083

Brand: Toronto Research Chemicals TRCI873000100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl-beta-D-thiogalactopyranoside
CAS 367-93-1
Molecular Formula C9 H18 O5 S
Quantity 100 g
Synonym Isopropyl -B-D-Thiogalactopyranoside, Dioxane Free
InChI Formula InChI=1S/C9H18O5S/c1-4(2)15-9-8(13)7(12)6(11)5(3-10)14-9/h4-13H,3H2,1-2H3/t5-,6+,7+,8-,9+/m1/s1
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
Molecular Weight (g/mol) 238.3
Formula Weight 238.0875
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.