missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl cyanoacetate, 97%

Product Code. 11413864
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413864 100 g 100g
11403864 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413864 Supplier Thermo Scientific Alfa Aesar Supplier No. A11887.22
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13361-30-3 | C6H9NO2 | 127.143 g/mol

Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing KnÖevenagel reactions with aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing Knöevenagel reactions with aldehydes and ketones.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13361-30-3
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00019842
InChI Key BESQLCCRQYTQQI-UHFFFAOYSA-N
Synonym isopropyl cyanoacetate, isopropyl 2-cyanoacetate, cyanoacetic acid isopropyl ester, acetic acid, cyano-, isopropyl ester, unii-0v3mgi9xma, 0v3mgi9xma, acetic acid, cyano-, 1-methylethyl ester, propan-2-yl cyanoacetate, prop-2-yl cyanoacetate, acmc-1bsph
PubChem CID 25917
IUPAC Name propan-2-yl 2-cyanoacetate
SMILES CC(C)OC(=O)CC#N

Specifications

Density 1.013
Boiling Point 207°C
Flash Point 92°C (198°F)
Refractive Index 1.415
Quantity 100 g
Beilstein 1756014
Solubility Information Slightly soluble in water.
Formula Weight 127.14
Percent Purity 97%
Chemical Name or Material Isopropyl cyanoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.