missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl chloroacetate, 99%

Product Code. 11468860
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468860 25 mL 25mL
11499366 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468860 Supplier Thermo Scientific Alfa Aesar Supplier No. A19070.AC
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 105-48-6 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-48-6
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00040410
InChI Key VODRWDBLLGYRJT-UHFFFAOYSA-N
Synonym isopropyl chloroacetate, chloroacetic acid isopropyl ester, acetic acid, chloro-, 1-methylethyl ester, iso-propyl chloroacetate, clch2c o och ch3 2, ccris 7748, acetic acid, chloro-, isopropyl ester, acetic acid, 2-chloro-, 1-methylethyl ester, chloroacetic acid, 1-methyl ester, monochloroacetic acid isopropyl ester
PubChem CID 7759
IUPAC Name propan-2-yl 2-chloroacetate
SMILES CC(C)OC(=O)CCl

Specifications

Density 1.08
Boiling Point 149°C to 150°C
Linear Formula ClCH2CO2CH(CH3)2
Refractive Index 1.4205
Quantity 25 mL
UN Number UN2947
Formula Weight 136.58
Percent Purity 99%
Chemical Name or Material Isopropyl chloroacetate
compliance-icons compliance-icons
Product Identifier
  • Isopropyl chloroacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Isopropyl chloroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.