missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free

Product Code. p-151655 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10031991

Brand: Thermo Scientific Chemicals 302790010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 367-93-1 | C9H18O5S | 238.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
PubChem CID 656894
ChEBI CHEBI:61448
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 17, IV, 3726
Merck Index 15, 5126
Specific Rotation Condition − 31.00 (20.00°C c=1,water)
Specific Rotation -29° to -33° (20°C, 589nm) (c=1, water)
Formula Weight 238.31
Percent Purity ≥98.5%
Grade Biochemical
Physical Form Powder or Flakes
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.