missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, Sterile, Made with USP Water for Injection, 70% Solution, BAKER™, J.T. Baker™
338.20 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 gL
Packaging:
Poly Bottle
Specifications
CAS | 67-63-0 |
Molecular Formula | C3H8O |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
SMILES | CC(C)O |
IUPAC Name | propan-2-ol |
Molecular Weight (g/mol) | 60.096 |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
Formula Weight | 60.1 |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'