missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, AR™, Macron Fine Chemicals™

Product Code. 15518744
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15518744 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518744 Supplier Macron Fine Chemicals Supplier No. 677525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Analytical grade solvent with a combination of value and purity for general laboratory and stockroom use.

AR™ grade solvents from Macron Fine Chemicals™ deliver reliable and consistent results to laboratories the world over. These solvents meet or exceed general laboratory standards for analytical grade products, helping ensure the integrity of results in applications.

  • Quality that ensures consistent results
  • Specifications that meet or exceed AR grade requirements
  • Produced in ISO certified facilities to ensure reliability

Specifications

Melting Point -88.5°C
CAS 67-63-0
Color Colorless
Quantity 2.5 L
Flash Point 12°C
Odor Rubbing alcohol
Physical Form Liquid
Residue after Evaporation ≤0.001%
Titratable Acid ≤0.0005meq/g
Molecular Formula C3H8O
Titratable Base ≤0.0001meq/g
Vapor Pressure 6.0 kPa
Water ≤0.1%
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Boiling Point 82°C
Solubility Information Miscible in water
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.10g/mol
Percent Purity ≥99.7%
Chemical Name or Material Isopropyl alcohol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5LT ISOPROPYL ALCOHOL AR .
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.