missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetoacetate-d7, TRC

Product Code. 30393887
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393887 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30393887 Supplier Toronto Research Chemicals Supplier No. TRCI82372710G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl Acetoacetate-d7
CAS 1329502-96-6
Molecular Formula C7H5D7O3
Quantity 10 g
Synonym 3-Oxo-butanoic Acid 1-Methylethyl Ester-d7, 3-Oxobutanoic Acid Isopropyl Ester-d7, Acetoacetic Acid Isopropyl Ester-d7, Isopropyl Acetoacetate-d7
InChI Formula InChI=1S/C7H12O3/c1-5(2)10-7(9)4-6(3)8/h5H,4H2,1-3H3/i1D3,2D3,5D
SMILES [2H]C([2H])([2H])C([2H])(OC(=O)CC(=O)C)C([2H])([2H])[2H]
IUPAC Name 1,1,1,2,3,3,3-heptadeuteriopropan-2-yl 3-oxobutanoate
Molecular Weight (g/mol) 151.21
Formula Weight 151.1226
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.