missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetate-d10, TRC

Product Code. 30399696
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399696 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30399696 Supplier Toronto Research Chemicals Supplier No. TRCI823417100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl Acetate-d10
CAS 1398065-53-6
Molecular Formula C5 D10 O2
Quantity 100 mg
Synonym Acetic Acid Isopropyl Ester-D10, 1-Methylethyl acetate-d10, 2-Acetoxypropane-d1, 2-Propyl-1,1,1,2,3,3,3-d7 acetate-2,2,2-d3, Isopropyl-d7 acetate-d3, Isopropyl acetate-d10, Isopropyl-d3 ethanoate-d3, iso-Propyl Acetate-d10
InChI Formula InChI=1S/C5H10O2/c1-4(2)7-5(3)6/h4H,1-3H3/i1D3,2D3,3D3,4D
SMILES [2H]C([2H])([2H])C(=O)OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name 1,1,1,2,3,3,3-heptadeuteriopropan-2-yl 2,2,2-trideuterioacetate
Molecular Weight (g/mol) 112.193
Formula Weight 112.131
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.