missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetate, 98%, Honeywell™

Product Code. 15661040 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15661040 1 L Glass Bottle 1 L
15671040 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15661040 Supplier Honeywell Research Chemicals Supplier No. 1129921L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.98

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-21-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008877
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Synonym isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C

Specifications

Melting Point -73°C
Color Colorless
Density 0.872g/mL at 25°C
Boiling Point 85°C
Explosion Limit 1.8% (37°F), 8%
Flash Point 4°C (39.2°F)
Ignition Point 894°F
Packaging Glass Bottle
Linear Formula CH3COOCH(CH3)2
Refractive Index n20/D 1.377(lit.)
Quantity 1 L
UN Number UN1220
Beilstein 1740761
Vapor Density 3.5 (vs air)
Formula Weight 102.13g/mol
Vapor Pressure 47mmHg (20°C)
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isopropyl Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.