missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 5-(Diphenylphosphoryl)pentanoate, TRC

Product Code. 30391459
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391459 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391459 Supplier Toronto Research Chemicals Supplier No. TRCI824450100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl 5-(Diphenylphosphoryl)pentanoate
CAS 2088449-88-9
Purity Grade Notes HPLC
Molecular Formula C20H25O3P
Quantity 100 mg
Synonym Isopropyl 5-(Diphenylphosphinyl)pentanoic Acid
InChI Formula InChI=1S/C20H25O3P/c1-17(2)23-20(21)15-9-10-16-24(22,18-11-5-3-6-12-18)19-13-7-4-8-14-19/h3-8,11-14,17H,9-10,15-16H2,1-2H3
SMILES CC(C)OC(=O)CCCCP(=O)(c1ccccc1)c2ccccc2
IUPAC Name isopropyl 5-diphenylphosphorylpentanoate
Molecular Weight (g/mol) 344.38
Formula Weight 344.154
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.