missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-Hydroxy-3,5-diisopropylbenzoate, TRC

Product Code. 30393754
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393754 250 mg -
30395093 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393754 Supplier Toronto Research Chemicals Supplier No. TRCI822250250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2095678-97-8
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.36
Synonym 4-Hydroxy-3,5-bis(1-methylethyl), Benzoic Acid 1-Methylethyl Ester
SMILES CC(C)OC(=O)c1cc(C(C)C)c(O)c(c1)C(C)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H24O3/c1-9(2)13-7-12(16(18)19-11(5)6)8-14(10(3)4)15(13)17/h7-11,17H,1-6H3
Chemical Name or Material Isopropyl 4-Hydroxy-3,5-diisopropylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.