missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-Aminobenzoate, TRC

Product Code. 30398606
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398606 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398606 Supplier Toronto Research Chemicals Supplier No. TRCI9006555G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl 4-Aminobenzoate
CAS 18144-43-9
Molecular Formula C10 H13 N O2
Quantity 5 g
Synonym Benzoic acid, 4-amino-, 1-methylethyl ester, Benzoic acid, p-amino-, isopropyl ester (6CI,7CI,8CI), 1-Methylethyl 4-aminobenzoate, Isopropyl 4-aminobenzoate, Isopropyl p-aminobenzoate, 4-Aminobenzoic acid 1-methylethyl ester
InChI Formula InChI=1S/C10H13NO2/c1-7(2)13-10(12)8-3-5-9(11)6-4-8/h3-7H,11H2,1-2H3
SMILES CC(C)OC(=O)c1ccc(N)cc1
IUPAC Name propan-2-yl 4-aminobenzoate
Molecular Weight (g/mol) 179.22
Formula Weight 179.0946
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.