missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-Aminobenzoate, Mikromol™

Product Code. 30051961
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051961 100 mg 100mg
30049182 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051961 Supplier Mikromol Supplier No. MM1255.08
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Isopropyl 4-Aminobenzoate
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Isopropyl 4-Aminobenzoate
CAS 18144-43-9
Analyte or Component Names Isopropyl 4-Aminobenzoate
Shipping Condition Room Temperature
Molecular Formula C10 H13 N O2
Certifications/Compliance ISO 17025
Synonym Isopropyl 4-Aminobenzoate, 4-Aminobenzoic Acid 1-Methylethyl Ester, Benzoic acid, p-amino-, isopropyl ester (6CI,7CI,8CI), 1-Methylethyl 4-aminobenzoate, Isopropyl 4-aminobenzoate, Isopropyl p-aminobenzoate
InChI Formula InChI=1S/C10H13NO2/c1-7(2)13-10(12)8-3-5-9(11)6-4-8/h3-7H,11H2,1-2H3
SMILES CC(C)OC(=O)c1ccc(N)cc1
IUPAC Name propan-2-yl 4-aminobenzoate
Molecular Weight (g/mol) 179.22
Quantity 100 mg
Formula Weight 179.0946 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.