missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4,4,4-trifluoroacetoacetate, 95%, Thermo Scientific™

Product Code. 11398285
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398285 25 g 25g
11308295 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398285 Supplier Thermo Scientific Alfa Aesar Supplier No. L10193.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175230-50-9
Molecular Formula C7H9F3O3
Molecular Weight (g/mol) 198.14
MDL Number MFCD00040990
InChI Key XLBGYZLICFMDDS-UHFFFAOYSA-N
Synonym isopropyl 4,4,4-trifluoroacetoacetate, isopropyl 4,4,4-trifluoro-3-oxobutanoate, isopropyl 4,4,4-trifluoro-3-oxobutyrate, isopropyl-4,4,4-trifluoroacetoacetate, acmc-209e9i, isopropyl4,4,4-trifluoroacetoacetate, methylethyl 4,4,4-trifluoro-3-oxobutanoate, isopropyl 4,4,4-trifluoro-3-oxobutanoate #, 4,4,4-trifluoroacetoacetic acid isopropyl ester
PubChem CID 586897
IUPAC Name propan-2-yl 4,4,4-trifluoro-3-oxobutanoate
SMILES CC(C)OC(=O)CC(=O)C(F)(F)F

Specifications

Boiling Point 42°C (28 mmHg)
Refractive Index 1.3891
Quantity 25 g
Beilstein 7475563
Formula Weight 198.14
Percent Purity 95%
Chemical Name or Material Isopropyl 4,4,4-trifluoroacetoacetate
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.