missing translation for 'onlineSavingsMsg'
Learn More

isopropyl 2-bromobenzoate, Thermo Scientific™

Product Code. 10165892
Click to view available options
Quantity:
1 g
10 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 59247-52-8
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
InChI Key QTCRCHHNKWSPHB-UHFFFAOYSA-N
PubChem CID 603261
IUPAC Name propan-2-yl 2-bromobenzoate
SMILES CC(C)OC(=O)C1=CC=CC=C1Br

Specifications

Assay Percent Range 97+%
Quantity 1 g
Synonym isopropyl 2-bromobenzoate, isopropyl o-bromobenzoate, methylethyl 2-bromobenzoate, prop-2-yl 2-bromobenzoate, isopropyl 2-bromobenzoate #, isopropyl2-bromobenzoate, 2-bromobenzoic acid isopropyl ester, 2-bromobenzoic acid, isopropyl ester, benzoic acid, 2-bromo-,1-methylethyl ester, benzoic acid, 2-bromo-, 1-methylethyl ester
Formula Weight 243.1
Percent Purity 97%
Chemical Name or Material Isopropyl 2-bromobenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'