missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 2-bromo-2-methylpropanoate, 97%, Thermo Scientific™

Product Code. p-2704613
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
This item is not returnable. View return policy

Product Code. 10160792

Brand: Thermo Scientific Maybridge SB01764EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 51368-55-9
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00055235
InChI Key UNZJYKKJZGIFCG-UHFFFAOYSA-N
Synonym isopropyl 2-bromo-2-methylpropanoate, isopropyl 2-bromo-2-methylpropionate, 2-bromo-2-methylpropionic acid isopropyl ester, isopropyl 2-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, 1-methylethyl ester, isopropyl 2-bromo-2-methyl-propanoate, methylethyl 2-bromo-2-methylpropanoate, 2-bromo-iso-butyric acid isopropyl ester, propan-2-yl 2-bromanyl-2-methyl-propanoate, 2-bromoisobutyric acid isopropyl ester
PubChem CID 2796293
IUPAC Name propan-2-yl 2-bromo-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)Br

Specifications

CAS Min % 97.0
Density 1.2600g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula (CH3)2CBrCO2CH(CH3)2
Quantity 10 g
Formula Weight 209.08
Percent Purity 97%
Chemical Name or Material Isopropyl 2-bromo-2-methylpropanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.