missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl 2-bromo-2-methylpropanoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
CAS Min % | 97.0 |
Density | 1.2600g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | (CH3)2CBrCO2CH(CH3)2 |
Quantity | 10 g |
Formula Weight | 209.08 |
Percent Purity | 97% |
Chemical Name or Material | Isopropyl 2-bromo-2-methylpropanoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'