missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl (1R)-(+)-Camphorsulfate, TRC

Product Code. 30399829
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399829 1 g 1g
30389248 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399829 Supplier Toronto Research Chemicals Supplier No. TRCI8243751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1242184-40-2
Molecular Formula C13 H22 O4 S
Molecular Weight (g/mol) 274.38
Synonym 1-Methylethyl (1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptane-1-methanesulfonate
IUPAC Name isopropyl [(1R,4S)-7,7-dimethyl-2-oxo-norbornan-1-yl]methanesulfonate
SMILES C(S(OC(C)C)(=O)=O)[C@]12C(C)(C)[C@](CC1=O)(CC2)[H]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H22O4S/c1-9(2)17-18(15,16)8-13-6-5-10(7-11(13)14)12(13,3)4/h9-10H,5-8H2,1-4H3/t10-,13-/m0/s1
Formula Weight 274.1239
Percent Purity >95%
Chemical Name or Material Isopropyl (1R)-(+)-Camphorsulfate
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.