missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 1-(1-phenylethyl)-1H-imidazole-5-carboxylate, TRC

Product Code. 30406380
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406380 100 mg -
30401282 50 mg 50mg
30404061 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406380 Supplier Toronto Research Chemicals Supplier No. TRCI485580100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 792842-51-4
Molecular Formula C15H18N2O2
Molecular Weight (g/mol) 258.32
Synonym 1-(1-Phenylethyl)-1H-Imidazole-5-carboxylic Acid Methylethyl EsterDesethyl-isopropyl Etomidate
IUPAC Name isopropyl 1-(1-phenylethyl)-1H-imidazole-5-carboxylate
SMILES CC(C)OC(=O)c1cncn1C(C)c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H18N2O2/c1-11(2)19-15(18)14-9-16-10-17(14)12(3)13-7-5-4-6-8-13/h4-12H,1-3H3
Formula Weight 258.14
Percent Purity >95%
Chemical Name or Material Isopropyl 1-(1-phenylethyl)-1H-imidazole-5-carboxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.