missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 1-(1-Phenylethyl)-1H-imidazole-5-carboxylate (1mg/mL in DMSO), TRC

Product Code. 30396320
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396320 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396320 Supplier Toronto Research Chemicals Supplier No. TRCKIT41681X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl 1-(1-phenylethyl)-1H-imidazole-5-carboxylate
CAS 792842-51-4
Molecular Formula C15H18N2O2
Quantity 1x1 mL
Synonym 1-(1-Phenylethyl)-1H-Imidazole-5-carboxylic Acid Methylethyl EsterDesethyl-isopropyl Etomidate
InChI Formula InChI=1S/C15H18N2O2/c1-11(2)19-15(18)14-9-16-10-17(14)12(3)13-7-5-4-6-8-13/h4-12H,1-3H3
SMILES CC(C)OC(=O)c1cncn1C(C)c2ccccc2
IUPAC Name isopropyl 1-(1-phenylethyl)-1H-imidazole-5-carboxylate
Molecular Weight (g/mol) 258.32
Formula Weight 258.14
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.