missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyl acetate, 99%

Product Code. 11487497
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487497 100 mL 100mL
11497497 500 mL 500mL
11407507 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487497 Supplier Thermo Scientific Alfa Aesar Supplier No. A15618.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-22-5 | C5H8O2 | 100.12 g/mol

Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols, phenols and amines under solvent free conditions. It is used as a solvent for cellulose, plastics, oil and fats. It is also a building block used for various organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols, phenols and amines under solvent free conditions. It is used as a solvent for cellulose, plastics, oil and fats. It is also a building block used for various organic synthesis.

Solubility
Slightly soluble in water. Soluble in alcohol and propylene glycol, acetone and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-22-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008709
InChI Key HETCEOQFVDFGSY-UHFFFAOYSA-N
Synonym isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate
PubChem CID 7916
IUPAC Name prop-1-en-2-yl acetate
SMILES CC(=C)OC(C)=O

Specifications

Melting Point -93°C
Boiling Point 93°C to 94°C
Flash Point 18°C (64°F)
Quantity 100 mL
UN Number UN2403
Beilstein 1280347
Merck Index 14,5203
Formula Weight 100.12
Percent Purity 99%
Chemical Name or Material Isopropenyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.