missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Molecular Biology Grade, Fisher BioReagents

For nucleic acid precipitation

Brand:  Fisher Bioreagents BP2618-212

Code : 32

Additional Details : CAS Number : 67-63-0 Weight : 3.20000kg Transport : UN number : 1219 Chem class : 1 Pack group : II


Quantity 2.5L
Packaging Glass Bottle

 View more versions of this product

Product Code. 11358461

  • £50.57 / 2.50 L


In Stock 11
EU Stock 3
Add to basket


Specifications

Chemical Identifiers

Isopropanol
Molecular Biology Grade
≥99.9%
C3H8O
60.1
67-63-0
DNase-, RNase- and Protease-Free
≥99.9 %
(CH3)2CHOH
2-Propanol

Specifications

Isopropanol
67-63-0
Vapor Pressure: 43mmHg at 20°C
DNase-, RNase- and Protease-Free
≥99.9%
C3H8O
60.1
Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids and proteins and preservation of biological specimens.
DNase free
Glass Bottle
0.002% max.
1ppm max.
Pass Test
43mmHg at 20°C
2.27 mPa/s at 20°C
Colorless
7
0.015 AU max. at 254nm
0.002% max.
Molecular Biology Grade
Liquid
≥99.9 %
(CH3)2CHOH
2-Propanol
0.785g/cm³
1 ppb max.
Protease free
1.3740 to 1.3760 (at 25°C)
Pass Test
0.0001mEq/g. max.
0.05% max.
81°C
−89.5°C
2.5L
Videos
SDS Documentation