missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, HPLC for Gradient Analysis, Fisher Chemical™

Product Code. 10561802 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10561802 2.5 L Amber Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10561802 Supplier Fisher Chemical Supplier No. P/7508/17
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

C3H8O, CAS Number-67-63-0, 2-hydroxypropane, 2-propanol, alkolave, avantine, dimethylcarbinol, hartosol, isopropanol, isopropyl alcohol, petrohol, sec-propyl alcohol, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096 | CAS: 67-63-0 | C3H8O | 60.096 g/mol

C3H8O, CAS Number-67-63-0, 2-hydroxypropane, 2-propanol, alkolave, avantine, dimethylcarbinol, hartosol, isopropanol, isopropyl alcohol, petrohol, sec-propyl alcohol, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096

TRUSTED_SUSTAINABILITY

Specifications

pH 7
Melting Point -89.5°C
CAS 67-63-0
Color Colorless
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula C3H8O
Vapor Pressure 43mmHg at 20°C
MDL Number 11674
Viscosity 2.27 mPaS at 20°C
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Boiling Point 81°C
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.1g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Propan-2-ol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.