missing translation for 'onlineSavingsMsg'
Learn More

Isoprocarb, TRC

Product Code. 30005890
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005890 1 g 1g
30135844 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005890 Supplier Toronto Research Chemicals Supplier No. TRCI8224001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2631-40-5
Molecular Formula C11 H15 N O2
Molecular Weight (g/mol) 193.24
Synonym Isoprocarb, Carbamic acid, methyl-, o-cumenyl ester (7CI,8CI), Phenol, 2-(1-methylethyl)-, methylcarbamate (9CI), 2-Isopropylphenyl N-methylcarbamate, 2-Isopropylphenyl methylcarbamate, BAY 39731, Bayer 39,731, ENT 25670, Etrofolan, Isoprocarb, MIPC, Mipcin, Mipcine, Mobucin, NSC 191479, OMS 32, PPC 3, Ro 7-5050, o-Cumenyl N-methylcarbamate, o-Cumenyl methylcarbamate, o-Isopropylphenol methylcarbamate, o-Isopropylphenyl N-methylcarbamate, o-Isopropylphenyl methylcarbamate, Phenol, 2-(1-methylethyl)-, 1-(N-methylcarbamate)
IUPAC Name (2-propan-2-ylphenyl) N-methylcarbamate
SMILES CNC(=O)Oc1ccccc1C(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H15NO2/c1-8(2)9-6-4-5-7-10(9)14-11(13)12-3/h4-8H,1-3H3,(H,12,13)
Formula Weight 193.1103
Chemical Name or Material Isoprocarb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.