missing translation for 'onlineSavingsMsg'
Learn More

Isoprene (Stabilized with TBC), TRC

Product Code. 30402944
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402944 10 g 10g
30403863 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402944 Supplier Toronto Research Chemicals Supplier No. TRCI82230010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-79-5
Molecular Formula C5 H8
Molecular Weight (g/mol) 68.12
Synonym 1,3-Butadiene, 2-methyl-, Isoprene (8CI), 2-Methyl-1,3-butadiene, 2-Methylbutadiene, 3-Methyl-1,3-butadiene, Isopentadiene, NSC 9237, β-Methylbivinyl
IUPAC Name 2-methylbuta-1,3-diene
SMILES CC(=C)C=C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H8/c1-4-5(2)3/h4H,1-2H2,3H3
Formula Weight 68.0626
Chemical Name or Material Isoprene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.