missing translation for 'onlineSavingsMsg'
Learn More

Isoprene monoxide, 95%

Product Code. 11359715
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359715 25 g 25g
11349715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359715 Supplier Thermo Scientific Alfa Aesar Supplier No. L19768.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1838-94-4 | C5H8O | 84.118 g/mol

Isoprene monoxide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoprene monoxide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1838-94-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00075225
InChI Key FVCDMHWSPLRYAB-UHFFFAOYSA-N
Synonym 2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 4,4,4-3fmet-1-2-furyl-1,3-butandione
PubChem CID 92166
IUPAC Name 2-ethenyl-2-methyloxirane
SMILES CC1(CO1)C=C

Specifications

Density 0.857
Boiling Point 79°C to 81°C
Flash Point −8°C (17°F)
Odor Recognizable
Refractive Index 1.416
Quantity 25 g
UN Number UN1993
Beilstein 104216
Solubility Information Slightly soluble in water.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material Isoprene monoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.