missing translation for 'onlineSavingsMsg'
Learn More

Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol

Product Code. 11373616
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373616 500 mL 500mL
11363616 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373616 Supplier Thermo Scientific Alfa Aesar Supplier No. L14619.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 78-79-5 | C5H8 | 68.119 g/mol

Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.

Solubility
Miscible with ethanol, ethyl ether, alcohol, acetone and benzene. Immiscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, ammonia, alkali metals, alcohols and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-79-5
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00008600
InChI Key RRHGJUQNOFWUDK-UHFFFAOYSA-N
Synonym isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
PubChem CID 6557
ChEBI CHEBI:35194
IUPAC Name 2-methylbuta-1,3-diene
SMILES CC(=C)C=C

Specifications

Melting Point -146°C
Density 0.681
Boiling Point 33°C to 36°C
Flash Point −54°C (−65°F)
Refractive Index 1.422
Quantity 500 mL
UN Number UN1218
Beilstein 969158
Merck Index 14,5201
Solubility Information Miscible with ethanol,ethyl ether,alcohol,acetone and benzene. Immiscible with water.
Formula Weight 68.12
Percent Purity 99%
Chemical Name or Material Isoprene, stabilized with ≈0.02% 4-tert-butylcatechol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isoprene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Isoprene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.