missing translation for 'onlineSavingsMsg'
Learn More

Isoprenaline hydrochloride, MedChemExpress

Product Code. 16441426
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
Product Code. Quantity unitSize
16441426 200 mg 200mg
16472189 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16441426

Brand: Medchem Express HYB0468

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isoprenaline hydrochloride is a non-selective β-adrenergic receptor agonist with potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities.

Isoprenaline hydrochloride is a non-selective β-adrenergic receptor agonist with potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities.

Specifications

Chemical Name or Material Isoprenaline hydrochloride
CAS 51-30-9
Color White
Purity Grade Notes Research
Molecular Formula C11H18ClNO3
Quantity 200 mg
Synonym Isoproterenol hydrochloride
Solubility Information DMSO : 80 mg/mL (322.95 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (201.84 mM)
SMILES OC1=CC=C(C(O)CNC(C)C)C=C1O.Cl
Molecular Weight (g/mol) 247.72
Formula Weight 247.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.