missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isoprenaline hemisulfate

Product Code. 15783457 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15783457 25 g 25g
15773457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15783457 Supplier Thermo Scientific Chemicals Supplier No. 462110250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 299-95-6 | C11H17NO3·1/2H2O4S | 520.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-95-6
Molecular Formula C11H17NO3·1/2H2O4S
Molecular Weight (g/mol) 520.59
InChI Key ZOLBALGTFCCTJF-UHFFFAOYSA-N
Synonym isoprenaline sulphate, isoprenaline sulfate, aleudrin, novodrine, aleudrine sulfate, isoproterenol sulfate, dl-isoprenaline sulfate, dl-isoproterenol sulfate, d,l-isoproterenol sulfate, +--isoprenaline sulfate
PubChem CID 8239
IUPAC Name 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;sulfuric acid
SMILES CC(C)NCC(C1=CC(=C(C=C1)O)O)O.CC(C)NCC(C1=CC(=C(C=C1)O)O)O.OS(=O)(=O)O

Specifications

Melting Point 83.0°C
Quantity 25 g
Formula Weight 520.59
Chemical Name or Material Isoprenaline hemisulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.