missing translation for 'onlineSavingsMsg'
Learn More

Merck Isopore™ Membrane Filters, 0.4 μm

Product Code. 10350181 Shop All Merck Products
Change view
Click to view available options
Pore Size:
0.4 μm
1.2 μm
Diameter (Metric):
13 mm
25 mm
37 mm
47 mm
90 mm
142 mm
Color:
Black
White
Unit Size:
Pack of 100
Pack of 30
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) Color unitSize
10350181 1.2 μm 13 mm White Pack of 100
10557682 0.4 μm 142 mm White Pack of 50
10752381 0.4 μm 13 mm White Pack of 100
10687891 0.4 μm 13 mm Black Pack of 100
10084500 0.4 μm 25 mm White Pack of 100
10074530 0.4 μm 25 mm Black Pack of 100
10116570 0.4 μm 37 mm White Pack of 100
10657301 0.4 μm 47 mm White Pack of 100
10339940 0.4 μm 47 mm Black Pack of 100
10024330 0.4 μm 90 mm White Pack of 30
10480221 1.2 μm 142 mm White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10350181 Supplier Merck Supplier No. RTTP01300
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Bubble Point 8.5 psi
Color White
Filter Type Screen Filter
Material Polycarbonate
Pore Size 1.2 μm
Sterility Non-sterile
Flow Rate Avg. 175 mL/min x cm2 (typical results @ 10 psi);687 mL/min x cm2/psi
For Use With (Application) Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence.
Gas Flow Rate 20.0 L/min/cm2
Hydrophobicity Hydrophilic
Quantity 100/Pk.
Type Filter Membranes
Wettability Hydrophilic
Diameter (Metric) 13 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.