Learn More
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Bubble Point | 50 psi |
| Color | White |
| Filter Type | Screen Filter |
| Material | Polycarbonate |
| Pore Size | 0.22 μm |
| Sterility | Non-sterile |
| Flow Rate | Avg. 16 mL/min x cm2 (typical results @ 10 psi);3.36 mL/min x cm2/psi |
| For Use With (Application) | Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence. |
| Gas Flow Rate | 1.0 L/min/cm2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.