missing translation for 'onlineSavingsMsg'
Learn More

Isopilocarpic Acid Sodium Salt, TRC

Product Code. 30132764
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132764 5 mg 5mg
30132415 25 mg 25mg
30041353 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132764 Supplier Toronto Research Chemicals Supplier No. TRCI8219605MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101769-87-3
Molecular Formula C11 H17 N2 O3 . Na
Molecular Weight (g/mol) 248.25
Synonym 1H-Imidazole-5-butanoic acid, α-ethyl-β-(hydroxymethyl)-1-methyl-, monosodium salt, [R-(R*,R*)]- (9CI), (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt, Pilocarpine Hydrochloride Imp. C (EP) as Sodium Salt, Isopilocarpic Acid Sodium Salt
IUPAC Name sodium;(2R,3R)-2-ethyl-3-(hydroxymethyl)-4-(3-methylimidazol-4-yl)butanoate
SMILES [Na+].CC[C@H]([C@H](CO)Cc1cncn1C)C(=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H18N2O3.Na/c1-3-10(11(15)16)8(6-14)4-9-5-12-7-13(9)2;/h5,7-8,10,14H,3-4,6H2,1-2H3,(H,15,16);/q;+1/p-1/t8-,10+;/m0./s1
Formula Weight 248.1137
Percent Purity >95%
Chemical Name or Material (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.