missing translation for 'onlineSavingsMsg'
Learn More

Isophthalaldehyde, 99%

Product Code. 10142960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142960 25 g Glass bottle 25g
10645202 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10142960 Supplier Thermo Scientific Acros Supplier No. 151740250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-19-7 | C8H6O2 | 134.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-19-7
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00003372
InChI Key IZALUMVGBVKPJD-UHFFFAOYSA-N
Synonym isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x
PubChem CID 34777
IUPAC Name benzene-1,3-dicarbaldehyde
SMILES C1=CC(=CC(=C1)C=O)C=O

Specifications

Melting Point 88°C to 90°C
Color White to Yellow
Boiling Point 136°C (17.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CHO)2
Quantity 25 g
Beilstein 07, 675
Solubility Information Solubility in water: slightly soluble
Formula Weight 134.13
Percent Purity 99%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material Isophthalaldehyde, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.